2-Fluoro-4-methylphenylboronic acid 97%

Warning

Synonyms: (2-Fluoro-4-methylphenyl)boronic acid2-Fluoro-4-methylbenzeneboronic acid

H53265-03 H53265-06
AAH53265-03EA 34.83 USD
AAH53265-03 AAH53265-06
2-Fluoro-4-methylphenylboronic acid 97%
2-Fluoro-4-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Melting Pt: 228…233 °C
Storage Temperature: Ambient
MDL Number: MFCD05664239
CAS Number: 170981-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR