4-(1H-Tetrazol-5-ylcarbamoyl)phenylboronic acid hydrochloride 97%

Warning

Synonyms: 4-(1H-Tetrazol-5-ylcarbamoyl)benzeneboronic acid hydrochloride(4-((1H-Tetrazol-5-yl)carbamoyl)phenyl)boronic acid hydrochloride

H53245-03 H53245-MD
AAH53245-03EA 279.12 USD
AAH53245-03 AAH53245-MD
4-(1H-Tetrazol-5-ylcarbamoyl)phenylboronic acid hydrochloride 97%
4-(1H-Tetrazol-5-ylcarbamoyl)phenylboronic acid hydrochloride
Formula: C₈H₉BClN₅O₃
MW: 269.45 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD06659882
CAS Number: 850568-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR