N-Isopropyl 3-boronobenzenesulfonamide 95%

Warning

Synonyms: 3-(Isopropylsulphamoyl)benzeneboronic acid3-(Isopropylsulfamoyl)benzeneboronic acid

H53236-MD H53236-03
AAH53236-MDEA 101.39 USD
AAH53236-MD AAH53236-03
N-Isopropyl 3-boronobenzenesulfonamide 95%
N-Isopropyl 3-boronobenzenesulphonamide
Formula: C₉H₁₄BNO₄S
MW: 243.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783856
CAS Number: 871329-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR