Potassium 4-(methylsulfonyl)phenyltrifluoroborate ≥96%

Warning

H53232-06 H53232-03
AAH53232-06EA 511.3 USD
AAH53232-06 AAH53232-03
Potassium 4-(methylsulfonyl)phenyltrifluoroborate ≥96%
Potassium 4-(methylsulphonyl)phenyltrifluoroborate
Formula: C₇H₇BF₃KO₂S
MW: 262.09 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD04115738
CAS Number: 850623-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR