2-Carboxythiophene-4-boronic acid 98%

Warning

H53210-MD
AAH53210-MDEA 140.73 USD
AAH53210-MD
2-Carboxythiophene-4-boronic acid 98%
2-Carboxythiophene-4-boronic acid
Formula: C₅H₅BO₄S
MW: 171.96 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD08689521
CAS Number: 913835-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR