(2-Methylsulfinylphenyl)boronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H53209-MD H53209-03
AAH53209-03EA 389.99 USD
AAH53209-03 AAH53209-MD
(2-Methylsulfinylphenyl)boronic acid 97%
(2-Methylsulfinylphenyl)boronic acid
Formula: C₇H₉BO₃S
MW: 184.02 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD03095261
CAS Number: 850567-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR