2,3,6-Trimethoxyphenylboronic acid ≥95%

Warning

Synonyms: (2,3,6-Trimethoxyphenyl)boronic acid2,3,6-Trimethoxybenzeneboronic acid

H53197-MD H53197-03
AAH53197-MDEA 49.54 USD
AAH53197-MD AAH53197-03
2,3,6-Trimethoxyphenylboronic acid ≥95%
2,3,6-Trimethoxyphenylboronic acid
Formula: C₉H₁₃BO₅
MW: 212.01 g/mol
Melting Pt: 34…40 °C
Storage Temperature: Ambient
MDL Number: MFCD03411944
CAS Number: 380430-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR