2-Aminophenylboronic acid pinacol ester hydrochloride 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Aminobenzeneboronic acid pinacol ester hydrochloride

H53194-03 H53194-MD
AAH53194-03EA 475.08 USD
AAH53194-03 AAH53194-MD
2-Aminophenylboronic acid pinacol ester hydrochloride 96%
2-Aminophenylboronic acid pinacol ester hydrochloride
Formula: C₁₂H₁₉BClNO₂
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD02179449
CAS Number: 393877-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR