4-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: 4-(3-Fluorophenylcarbamoyl)benzeneboronic acid(4-((3-Fluorophenyl)carbamoyl)phenyl)boronic acid

H53192-MD H53192-03
AAH53192-03EA 208.76 USD
AAH53192-03 AAH53192-MD
4-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%
4-(3-Fluorophenylcarbamoyl)phenylboronic acid
Formula: C₁₃H₁₁BFNO₃
MW: 259.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027204
CAS Number: 874288-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR