4-(Methoxycarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 4-(Methoxycarbamoyl)benzeneboronic acid(4-(Methoxycarbamoyl)phenyl)boronic acid

H53188-03 H53188-MD
AAH53188-MDEA 59.31 USD
AAH53188-MD AAH53188-03
4-(Methoxycarbamoyl)phenylboronic acid 97%
4-(Methoxycarbamoyl)phenylboronic acid
Formula: C₈H₁₀BNO₄
MW: 194.98 g/mol
Melting Pt: 130…136 °C
Storage Temperature: Ambient
MDL Number: MFCD04115670
CAS Number: 850568-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR