4-Chloro-3-(ethylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: 4-Chloro-3-(ethylcarbamoyl)benzeneboronic acid

H53175-03 H53175-MD
AAH53175-MDEA 119.62 USD
AAH53175-MD AAH53175-03
4-Chloro-3-(ethylcarbamoyl)phenylboronic acid 98%
4-Chloro-3-(ethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₁BClNO₃
Melting Pt: 220…224 °C
Storage Temperature: Ambient
MDL Number: MFCD07783861
CAS Number: 871332-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR