2-Isopropoxyphenylboronic acid 95%

Warning

Synonyms: 2-Isopropoxybenzeneboronic acid(2-Isopropoxyphenyl)boronic acid

H53168-06 H53168-03
AAH53168-03EA 43.14 USD
AAH53168-03 AAH53168-06
2-Isopropoxyphenylboronic acid 95%
2-Isopropoxyphenylboronic acid
Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
Boiling Pt: 199 °C
Melting Pt: 30…35 °C
Density: 1.075
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03427050
CAS Number: 138008-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR