3-Chloro-4-(ethylcarbamoyl)phenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(3-Chloro-4-(ethylcarbamoyl)phenyl)boronic acid
, 3-Chloro-4-(ethylcarbamoyl)benzeneboronic acid
H53162-03
H53162-MD
AAH53162-03EA
227.55
USD
AAH53162-03
AAH53162-MD
3-Chloro-4-(ethylcarbamoyl)phenylboronic acid 97%
3-Chloro-4-(ethylcarbamoyl)phenylboronic acid
Formula:
C₉H₁₁BClNO₃ MW: 227.45 g/mol Melting Pt: 142…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD07363767 CAS Number: 850589-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...