2-Methyl-5-nitrophenylboronic acid ≥96%

Warning

Synonyms: 2-Methyl-5-nitrobenzeneboronic acid

H53160-MD H53160-03
AAH53160-MDEA 125.53 USD
AAH53160-MD AAH53160-03
2-Methyl-5-nitrophenylboronic acid ≥96%
2-Methyl-5-nitrophenylboronic acid
Formula: C₇H₈BNO₄
MW: 180.96 g/mol
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00757435
CAS Number: 100960-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR