4-(2-Methoxyethylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: 4-(2-Methoxyethylcarbamoyl)benzeneboronic acid4-(2-Methoxyethylaminocarbonyl)phenylboronic acid

H53155-MD H53155-03
AAH53155-03EA 227.55 USD
AAH53155-03 AAH53155-MD
4-(2-Methoxyethylcarbamoyl)phenylboronic acid 98%
4-(2-Methoxyethylcarbamoyl)phenylboronic acid
Formula: C₁₀H₁₄BNO₄
MW: 223.04 g/mol
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD04115682
CAS Number: 850589-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR