(S)-2,2'-Dimethoxy-1,1'-binaphthalene-3,3'-diboronic acid 97%

Warning

Synonyms: (S)-2,2'-Dimethoxy-1,1'-binaphthyl-3,3'-diboronic acid

H53140-MD
AAH53140-MDEA 660.58 USD
AAH53140-MD
(S)-2,2'-Dimethoxy-1,1'-binaphthalene-3,3'-diboronic acid 97%
(S)-2,2'-Dimethoxy-1,1'-binaphthalene-3,3'-diboronic acid
Formula: C₂₂H₂₀B₂O₆
MW: 402.02
Storage Temperature: Ambient
MDL Number: MFCD09027249
CAS Number: 220204-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR