1-Methyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole1-Methyl-4-pyrazole boronic acid pinacol ester1-Methylpyrazole-4-boronic acid pinacol ester

H53139-03 H53139-MD
AAH53139-03EA 127.99 USD
AAH53139-03 AAH53139-MD
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₀H₁₇BN₂O₂
MW: 208.07 g/mol
Melting Pt: 59…64 °C
Storage Temperature: Ambient
MDL Number: MFCD03789259
CAS Number: 761446-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR