Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2,2,2-Trifluoroethoxy)phenylboronic acid 97%

Supplier: Alfa Aesar
Warning
Synonyms: 3-(2,2,2-Trifluoroethoxy)benzeneboronic acid

H53137-03 H53137-MD
AAH53137-03EA 212.18 USD InStock
AAH53137-03 AAH53137-MD
3-(2,2,2-Trifluoroethoxy)phenylboronic acid 97%
3-(2,2,2-Trifluoroethoxy)phenylboronic acid


Formula: C₈H₈BF₃O₃
MW: 219,95
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD07363846
CAS Number: 850593-08-7
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty