5-(Ethoxycarbonyl)-2-fluorophenylboronic acid 97%

Warning

Synonyms: (5-(Ethoxycarbonyl)-2-fluorophenyl)boronic acid5-Ethoxycarbonyl-2-fluorobenzeneboronic acid

H53134-MD H53134-03
AAH53134-03EA 254.41 USD
AAH53134-03 AAH53134-MD
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid 97%
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid
Formula: C₉H₁₀BFO₄
MW: 211.98 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD08235094
CAS Number: 874219-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR