3-Chloro-4-(dimethylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: 3-Chloro-4-(dimethylcarbamoyl)benzeneboronic acid(3-Chloro-4-(dimethylcarbamoyl)phenyl)boronic acid

H53131-03 H53131-MD
AAH53131-03EA 325.23 USD
AAH53131-03 AAH53131-MD
3-Chloro-4-(dimethylcarbamoyl)phenylboronic acid 98%
3-Chloro-4-(dimethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₁BClNO₃
MW: 227.45 g/mol
Melting Pt: 126…132 °C
Storage Temperature: Ambient
MDL Number: MFCD07363772
CAS Number: 850589-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR