4-Fluoro-(3-methoxycarbonyl)phenylboronic acid 98%

Warning

Synonyms: 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid4-Fluoro-3-methoxycarbonylphenylboronic acid

H53123-MD H53123-03
AAH53123-MDEA 32.61 USD
AAH53123-MD AAH53123-03
4-Fluoro-(3-methoxycarbonyl)phenylboronic acid 98%
4-Fluoro-(3-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BFO₄
MW: 197.96 g/mol
Melting Pt: 253…255 °C
Storage Temperature: Ambient
MDL Number: MFCD06801688
CAS Number: 874219-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR