4-(Methylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: 4-(N-Methylaminocarbonyl)phenylboronic acid4-(Methylcarbamoyl)benzeneboronic acid

H53090-03 H53090-MD
AAH53090-MDEA 57.06 USD
AAH53090-MD AAH53090-03
4-(Methylcarbamoyl)phenylboronic acid 98%
4-(Methylcarbamoyl)phenylboronic acid
Formula: C₈H₁₀BNO₃
MW: 178.98 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD03788427
CAS Number: 121177-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR