3-Fluoro-(5-methoxycarbonyl)phenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Fluoro-5-methoxycarbonylphenylboronic Acid3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid

H53084-03 H53084-MD
AAH53084-03EA 536.99 USD
AAH53084-03 AAH53084-MD
3-Fluoro-(5-methoxycarbonyl)phenylboronic acid 95%
3-Fluoro-(5-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BFO₄
MW: 197.96 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD07363749
CAS Number: 871329-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR