5-(tert-Butylcarbamoyl)pyridine-3-boronic acid 98%

Warning

H53081-03 H53081-MD
AAH53081-MDEA 123.21 USD
AAH53081-MD AAH53081-03
5-(tert-Butylcarbamoyl)pyridine-3-boronic acid 98%
5-(tert-Butylcarbamoyl)pyridine-3-boronic acid
Formula: C₁₀H₁₅BN₂O₃
MW: 222.05 g/mol
Melting Pt: 245…250 °C
Storage Temperature: Ambient
MDL Number: MFCD08689533
CAS Number: 913835-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR