5-Chloro-2-hydroxyphenylboronic acid 97%

Warning

Synonyms: 5-Chloro-2-hydroxybenzeneboronic acid

H53075-MD H53075-03
AAH53075-03EA 188.81 USD
AAH53075-03 AAH53075-MD
5-Chloro-2-hydroxyphenylboronic acid 97%
5-Chloro-2-hydroxyphenylboronic acid
Formula: C₆H₆BClO₃
MW: 172.37 g/mol
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD06659863
CAS Number: 89488-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR