2-Bromo-6-fluorophenylboronic acid 98%

Warning

Synonyms: 2-Bromo-6-fluorobenzeneboronic acid(2-Bromo-6-fluorophenyl)boronic acid

H53071-03 H53071-14 H53071-06
AAH53071-03EA 21.52 USD
AAH53071-03 AAH53071-14 AAH53071-06
2-Bromo-6-fluorophenylboronic acid 98%
2-Bromo-6-fluorophenylboronic acid
Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
Melting Pt: 142…145 °C
Storage Temperature: Ambient
MDL Number: MFCD05664309
CAS Number: 913835-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR