3-Chloro-4-(tert-butylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: 3-Chloro-4-(tert-butylcarbamoyl)benzeneboronic acid

H53069-03 H53069-MD
AAH53069-MDEA 81.27 USD
AAH53069-MD AAH53069-03
3-Chloro-4-(tert-butylcarbamoyl)phenylboronic acid 98%
3-Chloro-4-(tert-butylcarbamoyl)phenylboronic acid
Formula: C₁₁H₁₅BClNO₃
MW: 255.51
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD07363771
CAS Number: 850589-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR