2-Pyridineboronic acid dimethyl ester 95%

Supplier: Thermo Scientific Chemicals

Warning

H53067-03 H53067-MD
AAH53067-MDEA 144 USD
AAH53067-MD AAH53067-03
2-Pyridineboronic acid dimethyl ester 95%
2-Pyridineboronic acid dimethyl ester
Formula: C₇H₁₀BNO₂
MW: 150.97
Storage Temperature: Ambient
MDL Number: MFCD03412109
CAS Number: 136805-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR