4-(Heptyloxy)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 4-n-Heptyloxybenzeneboronic acid4-Heptyloxyphenylboronic acid4-Heptyloxybenzeneboronic acid

H53066-06 H53066-03
AAH53066-06EA 117 USD
AAH53066-06 AAH53066-03
4-(Heptyloxy)phenylboronic acid 97%
4-(Heptyloxy)phenylboronic acid
Formula: C₁₃H₂₁BO₃
MW: 236.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039169
CAS Number: 136370-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR