1-Boc-7-methoxyindole-2-boronic acid 98%

Warning

Synonyms: 7-Methoxy-1H-indole-2-boronic acid, N-BOC protected

H53063-03 H53063-MD
AAH53063-03EA 349.65 USD
AAH53063-03 AAH53063-MD
1-Boc-7-methoxyindole-2-boronic acid 98%
1-Boc-7-methoxyindole-2-boronic acid
Formula: C₁₄H₁₈BNO₅
MW: 291.11 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689542
CAS Number: 913835-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR