2-Carboxy-4-chlorophenylboronic acid 96%
|
Warning
|
Synonyms:
2-Carboxy-4-chlorobenzeneboronic acid
H53043-MD
H53043-03
AAH53043-MDEA
94.23
USD
AAH53043-MD
AAH53043-03
2-Carboxy-4-chlorophenylboronic acid 96%
2-Carboxy-4-chlorophenylboronic acid
Formula:
C₇H₆BClO₄ MW: 200.39 Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659869 CAS Number: 850568-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...