4-(Cyclopropylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 4-(Cyclopropylcarbamoyl)benzeneboronic acid

H53036-03 H53036-MD
AAH53036-03EA 132.44 USD
AAH53036-03 AAH53036-MD
4-(Cyclopropylcarbamoyl)phenylboronic acid 97%
4-(Cyclopropylcarbamoyl)phenylboronic acid
Formula: C₁₀H₁₂BNO₃
MW: 205.02 g/mol
Melting Pt: 202…206 °C
Storage Temperature: Ambient
MDL Number: MFCD03788419
CAS Number: 515140-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR