4-Fluor-3-(isopropylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: 4-Fluoro-3-(isopropylcarbamoyl)benzeneboronic acid(4-Fluoro-3-(isopropylcarbamoyl)phenyl)boronic acid

H53030-MD H53030-03
AAH53030-03EA 172.05 USD
AAH53030-03 AAH53030-MD
4-Fluor-3-(isopropylcarbamoyl)phenylboronic acid 98%
4-Fluor-3-(isopropylcarbamoyl)phenylboronic acid
Formula: C₁₀H₁₃BFNO₃
MW: 225.03 g/mol
Melting Pt: 263…265 °C
Storage Temperature: Ambient
MDL Number: MFCD08235040
CAS Number: 874219-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR