4-Chloro-3-nitrophenylboronic acid pinacol ester 98%

Synonyms: 2-(4-Chloro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane4-Chloro-3-nitrobenzeneboronic acid pinacol ester4-Chloro-3-nitrophenylboronic acid, pinacol ester

H53022-MD H53022-03
AAH53022-03EA 160.83 USD
AAH53022-03 AAH53022-MD
4-Chloro-3-nitrophenylboronic acid pinacol ester 98%
4-Chloro-3-nitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BClNO₄
MW: 283.52 g/mol
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD08235086
CAS Number: 913836-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR