5-Chloro-(2-ethoxycarbonyl)phenylboronic acid 98%
|
Warning
|
Synonyms:
(5-Chloro-2-(ethoxycarbonyl)phenyl)boronic acid
, 5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid
H53016-MD
H53016-03
AAH53016-MDEA
85.74
USD
AAH53016-MD
AAH53016-03
5-Chloro-(2-ethoxycarbonyl)phenylboronic acid 98%
5-Chloro-(2-ethoxycarbonyl)phenylboronic acid
Formula:
C₉H₁₀BClO₄ MW: 228.44 g/mol Melting Pt: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD07363744 CAS Number: 871329-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...