2-Chloro-(5-methoxycarbonyl)phenylboronic acid 98%

Warning

Synonyms: (2-Chloro-5-(methoxycarbonyl)phenyl)boronic acid2-Chloro-5-(methoxycarbonyl)benzeneboronic acid

H52999-03 H52999-MD
AAH52999-03EA 193.51 USD
AAH52999-03 AAH52999-MD
2-Chloro-(5-methoxycarbonyl)phenylboronic acid 98%
2-Chloro-(5-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BClO₄
MW: 214.41 g/mol
Melting Pt: 230…232 °C
Storage Temperature: Ambient
MDL Number: MFCD08689528
CAS Number: 913835-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR