5-Chloro-2-methylphenylboronic acid 97%

Warning

Synonyms: (5-Chloro-2-methylphenyl)boronic acid5-Chloro-2-methylbenzeneboronic acid

H52994-06 H52994-03
AAH52994-03EA 47.23 USD
AAH52994-03 AAH52994-06
5-Chloro-2-methylphenylboronic acid 97%
5-Chloro-2-methylphenylboronic acid
Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD03411939
CAS Number: 148839-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR