3-(Furfurylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 3-(Furfurylcarbamoyl)benzeneboronic acid

H52992-03 H52992-MD
AAH52992-03EA 170.45 USD
AAH52992-03 AAH52992-MD
3-(Furfurylcarbamoyl)phenylboronic acid 97%
3-(Furfurylcarbamoyl)phenylboronic acid
Formula: C₁₂H₁₂BNO₄
Melting Pt: 216…220 °C
Storage Temperature: Ambient
MDL Number: MFCD04115700
CAS Number: 850567-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR