4-(Diethylcarbamoyl)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(N,N-Diethylaminocarbonyl)phenylboronic acid4-(Diethylcarbamoyl)benzeneboronic acid

H52986-03 H52986-06
AAH52986-03EA 42.56 USD
AAH52986-03 AAH52986-06
4-(Diethylcarbamoyl)phenylboronic acid 98%
4-(Diethylcarbamoyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD03411949
CAS Number: 389621-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR