4-Fluoro-2-methoxyphenylboronic acid 98%
|
Warning
|
Synonyms:
4-Fluoro-2-methoxybenzeneboronic acid
H52976-03
H52976-06
AAH52976-06EA
203.66
USD
AAH52976-06
AAH52976-03
4-Fluoro-2-methoxyphenylboronic acid 98%
4-Fluoro-2-methoxyphenylboronic acid
Formula:
C₇H₈BFO₃ MW: 169.95 g/mol Melting Pt: 131…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788423 CAS Number: 179899-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...