2-Fluoro-(4-methoxycarbonyl)phenylboronic acid 98%

Warning

Synonyms: 2-Fluoro-4-(methoxycarbonyl)phenylboronic acid2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid

H52971-03 H52971-MD
AAH52971-MDEA 32.13 USD
AAH52971-MD AAH52971-03
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid 98%
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BFO₄
MW: 197.96 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD08436034
CAS Number: 603122-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR