3-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: 3-(3-Fluorophenylcarbamoyl)benzeneboronic acid

H52959-MD H52959-03
AAH52959-MDEA 49.08 USD
AAH52959-MD AAH52959-03
3-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%
3-(3-Fluorophenylcarbamoyl)phenylboronic acid
Formula: C₁₃H₁₁BFNO₃
MW: 259.04
Storage Temperature: Ambient
MDL Number: MFCD09027207
CAS Number: 874288-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR