4,4-Dimethylcyclohexene-1-ylboronic acid ≥96%

Warning

H52955-MD
AAH52955-MDEA 389.23 USD
AAH52955-MD
4,4-Dimethylcyclohexene-1-ylboronic acid ≥96%
4,4-Dimethylcyclohexene-1-ylboronic acid
Formula: C₈H₁₅BO₂
MW: 154.02
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD07363758
CAS Number: 865869-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR