4-Phenoxyphenylboronic acid 98%

Warning

Synonyms: (4-Phenoxyphenyl)boronic acid4-Phenoxybenzeneboronic acid

H52950-03 H52950-06
AAH52950-03EA 37.79 USD
AAH52950-03 AAH52950-06
4-Phenoxyphenylboronic acid 98%
4-Phenoxyphenylboronic acid
Formula: C₁₂H₁₁BO₃
MW: 214.03 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00093312
CAS Number: 51067-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR