4-Fluoro-3-(2-hydroxyethylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: 4-Fluoro-3-(2-hydroxyethylcarbamoyl)benzeneboronic acid

H52934-03 H52934-MD
AAH52934-MDEA 100.93 USD
AAH52934-MD AAH52934-03
4-Fluoro-3-(2-hydroxyethylcarbamoyl)phenylboronic acid 98%
4-Fluoro-3-(2-hydroxyethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₁BFNO₄
MW: 227.00
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD08235044
CAS Number: 874219-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR