3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Chloro-4-(cyclopropylcarbamoyl)phenyl)boronic acid3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid

H52910-MD H52910-03
AAH52910-03EA 285.27 USD
AAH52910-03 AAH52910-MD
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid 97%
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid
Formula: C₁₀H₁₁BClNO₃
MW: 239.46 g/mol
Melting Pt: 216…220 °C
Storage Temperature: Ambient
MDL Number: MFCD07363770
CAS Number: 850589-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR