2-Chloro-5-fluorophenylboronic acid 98%

Warning

Synonyms: 2-Chloro-5-fluorobenzeneboronic acid

H52906-06 H52906-03
AAH52906-06EA 496.08 USD
AAH52906-06 AAH52906-03
2-Chloro-5-fluorophenylboronic acid 98%
2-Chloro-5-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
MW: 174.37 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD06656272
CAS Number: 444666-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR