3-Bromo-5-(trifluoromethyl)phenylboronic acid 98%

Warning

Synonyms: 3-Bromo-5-(trifluoromethyl)benzeneboronic acid

H52898-03 H52898-MD
AAH52898-03EA 221.65 USD
AAH52898-03 AAH52898-MD
3-Bromo-5-(trifluoromethyl)phenylboronic acid 98%
3-Bromo-5-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BBrF₃O₂
MW: 268.82
Melting Pt: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD08436051
CAS Number: 913835-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR