4-tert-Butyl-3-nitrophenylboronic acid 98%

Warning

Synonyms: 4-tert-Butyl-3-nitrobenzeneboronic acid

H52883-03 H52883-MD
AAH52883-03EA 361.21 USD
AAH52883-03 AAH52883-MD
4-tert-Butyl-3-nitrophenylboronic acid 98%
4-tert-Butyl-3-nitrophenylboronic acid
Formula: C₁₀H₁₄BNO₄
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD07363743
CAS Number: 871329-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR