4-(Hexyloxy)phenylboronic acid 96%

Warning

Synonyms: B-[4-(Hexyloxy)phenyl]boronic acid4-n-Hexyloxybenzeneboronic acid

H52874-06 H52874-03
AAH52874-06EA 196.73 USD
AAH52874-06 AAH52874-03
4-(Hexyloxy)phenylboronic acid 96%
4-(Hexyloxy)phenylboronic acid
Formula: C₁₂H₁₉BO₃
MW: 222.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039027
CAS Number: 121219-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR